All Photos(1)

TMSTK-120ML

Sigma-Aldrich

TurboMix Stacking Solution, 120 mL

sterility

0.2 μm filtered

form

liquid

feed ratio

29:1

storage condition

protect from light

storage temp.

2-8°C

General description

TurboMix Stacking Solution is a 4% acrylamide buffer solution used to make bis-tris polyacrylamide gels. It is a ready-to-use solution for stacking gels. First, add APS and TEMED. After pouring the resolving gel, pour the stacking solution and insert your comb.

Application

Premixed stacking gel solution for casting bis-tris polyacrylamide gels
Bis-Tris Stacking Gel Solution

Legal Information

TURBOMIX is a trademark of Merck KGaA

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Articles

Bis Tris Polyacrylamide Gel Electrophoresis Technology

Bis-Tris gels and buffers for superior protein resolution compared to traditional tris-glycine gels.

Protocols

Hand-casting gels for PAGE and SDS-PAGE using TurboMix™ Bis-Tris Gel Casting Kits

Overview of polyacrylamide gel chemistry and detailed instructions for hand-casting polyacrylamide gels using TurboMix™ Bis-Tris Gel Casting Kits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service