Products
TW
EN
Products
Applications
Products
Services
Support
Home
Search Results
mPAGE
®
8-16% Bis-Tris Precast Gel
(3)
mPAGE
®
8-16% Bis-Tris Precast Gel
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
MP81G10
mPAGE
®
8-16% Bis-Tris Precast Gel, L × W × wells 10 cm × 8 cm × 10 well
Expand
View Pricing
MP81G12
mPAGE
®
8-16% Bis-Tris Precast Gel, L × W × wells 10 cm × 8 cm × 12 well
Expand
View Pricing
MP81G15
mPAGE
®
8-16% Bis-Tris Precast Gel, L × W × wells 10 cm × 8 cm × 15 well
Expand
View Pricing