Skip to Content
Merck
Titriplex® solution A for the determination of alkaline earth metals in water 1 ml ≙ 56 mg CaO/l using 100 ml of water, Titripur®

Titriplex® solution A

Titriplex® solution A for the determination of alkaline earth metals in water 1 ml ≙ 56 mg CaO/l using 100 ml of water, Titripur®
Synonym(s):
Hardness, determinations

Products

Product Category
Brand
Physical Form
Product Line
Quality Segment
Technique