跳轉至內容
Merck
NU075/441-321
NovAseptic® Tank outlet (3/4") Valve Body 90°
Valve Body - ASTM® A269/270 and Outlet with Elbow 90 °, Electro-polished with Butt end for welding
登入 查看組織和合約定價
選擇尺寸
關於此項目
分類程式碼代碼:
23151800
技術服務
需要援助?我們經驗豐富的科學家團隊隨時樂意為您提供支援。
讓我們為您提供幫助
技術服務
需要援助?我們經驗豐富的科學家團隊隨時樂意為您提供支援。
讓我們為您提供幫助
品質等級
參數
-1-10 bar max. pressure (-14.5-145 psig) at 25 °C (atmospheric pressure)
-80-200 °C max. temp. (-112-392 °F)
一般說明
NovAseptic® Valves are engineered for optimal performance, reliability, durability and ease of maintenance. NovAseptic® Valves comply with cGMP Design Qualification
criteria for aseptic processing. This is why we choose to call it ‘‘Aseptic by Design.”
應用
NovAseptic® Valves are specifically designed for aseptic applications and comply with the most stringent cleanability and sterilizability requirements. The focus on aseptic design is a significant feature for all valves. Flow through valve is a valve designed for a wide range of applications in the aseptic process; including manifolds, take off and drainage valves.
生化/生理作用
Surface roughness: Internal surface Ra ≤ 0.38 μm (15 μin) External surface Ra ≤ 1.6 μm (63 μin)
其他說明
All open ends on the valve body are protected with plastic covers on delivery and packaged in a closed box.
法律資訊
ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
分析證明 (COA)
輸入產品批次/批號來搜索 分析證明 (COA)。在產品’s標籤上找到批次和批號,寫有 ‘Lot’或‘Batch’.。
已經擁有該產品?
您可以在文件庫中找到最近購買的產品相關文件。
存取文件庫
我們的科學家團隊在所有研究領域都有豐富的經驗,包括生命科學、材料科學、化學合成、色譜、分析等.
聯絡技術服務