MilliporeSigma
Optimize your cell culture feed strategy
Maximize productivity with
Cellvento® ModiFeed Prime COMP
Development of upstream processes that maximize productivity without increasing complexity relies on the right combination of medium and feed and the ability to maintain tight control of culture conditions. In fed-batch mode, cells are initially grown in a relatively lean production medium followed by regular feed supplementation to achieve the desired product yield.
Cellvento® ModiFeed Prime COMP is a chemically-defined, highly concentrated, single-part, pH Neutral feed that allows for a reduction in volume of feed added to replenish depleted nutrients, thereby increasing product yield.
Use this new feed to reduce workflow complexity and risk to the process. Cellvento® ModiFeed Prime COMP is supplied as a compacted dry powder which reduces bulk volume, improves flowability, decreases dust formation, simplifies handling and accelerates hydration. Once hydrated, the medium can be stored protected from light at 2-8° C for up to 90 days or at room temperature for up to 30 days.
Take the right path upstream, with Cellvento® ModiFeed Prime COMP
Hydration and Stability Benefits. Other Suppliers vs. Cellvento ModiFeed Prime COMP
Products
Loading
Dry Powder Media vs. EZMix™ Compacted Media
Achieve easier handling of your feed
Request Free Cellvento® ModiFeed Prime COMP Sample
Thank you for your interest in the Cellvento® ModiFeed Prime COMP sample, reference 1.04132.0001.
To receive your free sample, please fill out and submit the form below.
ENTRY IN THIS PROMOTION CONSTITUTES YOUR ACCEPTANCE OF THESE OFFICIAL RULES.