Flavor & Fragrance Demo Kit Request
Thank you for your interest in our most recent F&F demo kit. Below is a list of products you will get to experience. After submitting the form, you will be re-directed to our website. We will be in touch with you soon in regards to your request.
| FEMA | Name | Organoleptics |
|---|---|---|
| 3403 | cis-3-Hexenyl hexanoate, natural, >=95%, FG | green; fruity; waxy; wine-like |
| 3505 | 3-Methylbutyl 2-methylbutanoate, natural, >=98%, FG | apple; blueberry; cherry; fruity; sweet |
| 3603 | 2-Octen-4-one, mixture of isomers, natural, >=97%, FG | green; mushroom; fruity; tropical |
| 3634 | 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, natural, >=97%, FG | caramel; maple; sweet |
| 3754 | Vanillin isobutyrate, natural, >=98%, FG | creamy; nutmeg; fruity; sweet; vanilla |
| 4170 | Methyl N-acetylanthranilate, >=99%, FG | floral; fruity |
| 4413 | trans-3-Hexenyl acetate, >=95%, stabilized, FG | banana; green; fruity; pear; rum |
| 4553 | cis-3-Nonenyl acetate, >=95%, stabilized, FG | green; herbaceous; pear; tropical |