Immobilon® Tools
for Western blotting
Western blotting success relies on having the right tools for the job. Our comprehensive Immobilon® portfolio offers what you need for dependable, reproducible, high sensitivity Western blotting.
Start with our selection of Immobilon® PVDF and nitrocellulose transfer membranes that allow you to choose the ideal membrane based on target protein size and detection method.
Immobilon® NOW dispensers and rolls offer the convenience of pre-cut sheets with the flexibility of rolls in the formats you depend on.
Highest sensitivity? Long signal duration? Immobilon® ECL (enhanced chemiluminescence) Western blotting HRP substrates were made to suit your needs.
Tired of immunodetection protocols that require hours of hands-on time? Accelerate immunodetection with the SNAP i.d.® 2.0 system for Western blotting. The SNAP i.d. 2.0 system uses a vacuum manifold to drive blocking reagents, antibodies, and wash buffers directly through the PVDF or nitrocellulose membrane, speeding immunodetection workflows.