Look Closer at Your Data
The xMAP® INTELLIFLEX System for Optimized Workflows
The Luminex® xMAP® INTELLIFLEX system is the latest addition to the xMAP® line of instruments. The compact xMAP® INTELLIFLEX system is a flow-based, multiplex platform that simplifies your workflow by combining the established performance of xMAP® technology with advanced features to elevate your assay performance and empower innovation in assay development. By combining low- and high-plex capabilities, the xMAP® INTELLIFLEX system gives fast, reliable results.
Stay up to date on the latest immunoassay research trends and our newest product offerings by subscribing to our Analyte Update newsletter or explore our current immunoassay offerings in the Analyte Quarterly.
Luminex® xMAP® INTELLIFLEX system combining established performance with advanced features.
xMAP® INTELLIFLEX System
Benefits of Our xMAP® Workflow With the New xMAP® INTELLIFLEX System
  • Extensive Assay Portfolio – Assay your way with MILLIPLEX® multiplex assays and xMAP® technology that offers the versatility to choose from the most extensive range of commercially available kits and customizable assays.
  • Wide Dynamic Range – Optimize your experiments by eliminating repeat testing from out-of-range values with the widest dynamic range of any xMAP® platform.
  • Smaller Size – Decrease space on your lab bench with a compact system that features an embedded PC and a touchscreen user interface.
  • User-Friendly, Flexible Software – Control the platform with easy-to-use instrument software that empowers you to quickly set up runs, optimize workflows, gather performance insights, and export data into common analysis tools.
  • Dual Reporter Functionality – Get more data from each target with the new second reporter channel that allows you to save on time and precious sample.
  • Easy Analysis – Analyze your data with our easily operated Belysa™ Immunoassay Curve Fitting software to optimize and compare standard curves, use rules-based data flagging, and confirm the integrity and reproducibility of data within a plate or across plates from multiple runs.
Request Info on xMAP® INTELLIFLEX System