MilliporeSigma
Mycosafe®: Mycoplasma Control for Quality Assurance
HomeMycosafe® Mycoplasma Reference Standards
Mycosafe® Mycoplasma Reference Standards
Validation Standards & Controls according to EP, USP, JP and ChP Requirements for Application in NAT- & Culture-based Mycoplasma Testing of 'Classical' Biopharmaceuticals and ATMPs in Pharma QC, Cell & Gene Therapies, and Regenerative Medicine
Mycoplasma pneumoniae FH Colonies 10 CFU
Mycosafe® Mycoplasma Reference Standards, developed by Professor Renate Rosengarten, are specifically designed for validating mycoplasma detection methods and for use as positive controls in routine mycoplasma testing of 'classical' biopharmaceuticals and cell and gene therapy products by ensuring compliance with regulatory requirements. These reference standard preparations therefore serve as a reliable tool in quality control and quality assurance to address the critical issue of mycoplasma contamination in biopharmaceutical production and cell culture systems.
Characterized by a low ratio of genome copies (GC) to the total mycoplasma cells (in colony-forming units, CFU) confirming the scientific accuracy of their production process, Mycosafe® Mycoplasma Reference Standards ensure an objective and unbiased comparability of NAT-based and culture-based mycoplasma detection methods, including Real-time PCR methods. Additionally, Mycosafe® Mycoplasma Reference Standards are offered not only as viable but also as heat-inactivated cell preparations, which provide a safe alternative for mycoplasma testing in environments where handling live mycoplasma cells may pose contamination risks.
As ready-to-use commercial products, Mycosafe® Mycoplasma Reference Standards streamline laboratory workflows and thus reduce contamination risks. They enable the implementation of rapid and reliable mycoplasma testing methods for biopharmaceutical and cell and gene therapy products, making them essential for maintaining product quality and ensuring patient safety in the rapidly evolving biopharmaceutical industry.
The Mycosafe® Mycoplasma Reference Standards will be available soon! Connect with our experts to get notified when they're available or to get more information!
Concentration of Total Mycoplasma Cells GC / CFU RatioMycoplasma SpeciesType StrainCulture CollectionCatalog Number – Mycosafe®
NCTCATCCNBRCViable
350 µL
 Heat Inactivated
350 µL
1000 CFU / 100 µL< 10Mycoplasma pneumoniaeFHNCTC 10119ATCC 15531NBRC 14401MP-T-1000MP-T-1000-HI
Mycoplasma oraleCH19299NCTC 10112ATCC 23714NBRC 14477MO-T-1000MO-T-1000-HI
Acholeplasma laidlawiiPG8NCTC 10116ATCC 23206NBRC 14400AL-T-1000AL-T-1000-HI
Mycoplasma hyorhinisBTS7NCTC 10130ATCC 17981NBRC 14858MH-T-1000MH-T-1000-HI
Mycoplasma fermentansPG18NCTC 10117ATCC 19989NBRC 14854MF-T-1000MF-T-1000-HI
Mycoplasma salivariumPG20NCTC 10113ATCC 23064NBRC 14478MSA-T-1000MSA-T-1000-HI
Mycoplasma argininiG230NCTC 10129ATCC 23838NBRC 111899MA-T-1000MA-T-1000-HI
Spiroplasma citriR8A2NCTC 10164ATCC 27556NA*SC-T-1000SC-T-1000-HI
Mycoplasma gallisepticumPG31NCTC 10115ATCC 19610NBRC 14855MG-T-1000MG-T-1000-HI
Mycoplasma synoviaeWVU 1853NCTC 10124ATCC 25204NA*MS-T-1000MS-T-1000-HI
NA*=Not Available
Request More Information
The Mycosafe® Mycoplasma Reference Standards will be available soon. Fill in the form below to get alerted when they are available or to request more information.
Fields indicated by an * are required.