Certificate of Analysis
Sigma-Aldrich
Product Name:
Phenyl acetate
Product Description:
99%
Product Brand:
Sigma-Aldrich
Product Number:
108723
Molecular Weight:
136.15
Molecular Formula:
CH3COOC6H5
CAS Number:
122-79-2
Test
Specification
Lot 03517AW Results
APPEARANCE:
COLORLESS LIQUID
COLORLESS LIQUID
REFRACTIVE INDEX AT:
1.5020
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE AND STANDARD AS
CONFORMS TO STRUCTURE AND STANDARD AS ILLUS-
 :
ILLUSTRATED ON PAGE 312B OF EDITION I,
TRATED ON PAGE 312B OF EDITION I, VOLUME 2
 :
VOLUME 2 OF "THE ALDRICH LIBRARY OF FT-IR
OF "THE ALDRICH LIBRARY OF FT-IR SPECTRA".
GAS LIQUID:
98.5% (MINIMUM)
99.6%
COLOR TEST:
50 APHA (MAXIMUM)
0<APHA<10
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.