Certificate of Analysis
Sigma-Aldrich
Product Name:
1,4-Butanediol divinyl ether
Product Description:
98%
Product Brand:
Sigma-Aldrich
Product Number:
123315
Molecular Weight:
142.20
Molecular Formula:
(-CH2CH2OCH=CH2)2
CAS Number:
3891-33-6
Test
Specification
Lot 09020JF Results
APPEARANCE:
COLORLESS LIQUID
COLORLESS LIQUID
REFRACTIVE INDEX AT:
1.4403
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE AND STANDARD AS
CONFORMS TO STRUCTURE AND STANDARD AS
 :
ILLUSTRATED ON PAGE 129A OF EDITION III OF
ILLUSTRATED ON PAGE 129A OF EDITION III OF
 :
"THE ALDRICH LIBRARY OF INFARED SPECTRA".
"THE ALDRICH LIBRARY OF INFRARED SPECTRA".
PROTON, CARBON NMR:
CONFORMS TO STRUCTURE.
GAS LIQUID:
97.5% (MINIMUM)
98.7%
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.