Certificate of Analysis
Sigma-Aldrich
Product Name:
Terbium(III) chloride hexahydrate
Product Description:
99.999% trace metals basis
Product Brand:
Sigma-Aldrich
Product Number:
204560
Molecular Weight:
373.38
Molecular Formula:
TbCl3 · 6H2O
CAS Number:
13798-24-8
Test
Specification
Lot 04124DH Results
APPEARANCE:
WHITE CRYSTALLINE POWDER AND/OR CHUNKS
WHITE CRYSTALLINE POWDER
TITRATION:
40.8% TB (MINIMUM) (COMPLEXOMETRIC)
42.9% TB (COMPLEXOMETRIC)
TRACE ANALYSIS, ICP-MS:
TRACE ELEMENTS IN PPM
 :
DY 2.2 CA 1.9 GD 1.6 NA 1.5 B 1.1
 :
BI 1.0 AL 0.9 IN 0.9 LI 0.8 YB 0.5
 :
ER 0.4 FE 0.4 EU 0.4 CU 0.3 LA 0.3
 :
PB 0.3 Y 0.2 HO 0.1 SM 0.1
ICP ASSAY:
CONFIRMS TERBIUM COMPONENT.
CONFIRMS TERBIUM COMPONENT
PURITY:
99.999% BASED ON RARE EARTH IMPURITIES
QUALITY CONTROL:
APRIL 2007
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.