Certificate of Analysis
Sigma-Aldrich
Product Name:
N-Methylbenzamide
Product Description:
≥99%
Product Brand:
Sigma-Aldrich
Product Number:
222798
Molecular Weight:
135.16
Molecular Formula:
C6H5CONHCH3
CAS Number:
613-93-4
Test
Specification
Lot 01021EZ Results
APPEARANCE:
WHITE TO OFF-WHITE POWDER, CRYSTALS OR CHIPS
WHITE CRYSTALLINE CHIPS
MELTING POINT:
80-82 DEGREES CELSIUS
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE
CONFORMS TO STRUCTURE AND STANDARD AS
 :
ILLUSTRATED ON PAGE 365D OF EDITION I,
 :
VOLUME 2 OF "THE ALDRICH LIBRARY OF FT-IR
 :
SPECTRA".
ELEMENTAL ANALYSIS:
CARBON 70.4%-71.8%
CARBON 71.24%
 :
NITROGEN 10.2%-10.5%
HYDROGEN 7.03%
 :
NITROGEN 10.46%
GAS LIQUID:
99.0% (MINIMUM)
99.8%
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.