Certificate of Analysis
Sigma-Aldrich
Product Name:
2,2,5,5-Tetramethyltetrahydrofuran
Product Description:
97%
Product Brand:
Sigma-Aldrich
Product Number:
223700
Molecular Weight:
128.21
CAS Number:
15045-43-9
Test
Specification
Lot 08707EU Results
APPEARANCE:
COLORLESS LIQUID
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE AND STANDARD AS
 :
ILLUSTRATED ON PAGE 377D OF EDITION I,
 :
VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR
 :
SPECTRA".
GAS LIQUID:
97.0 %
QUALITY CONTROL:
MAY 1999
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.