Certificate of Analysis
Sigma-Aldrich
Product Name:
3′-Fluoroacetanilide
Product Description:
98%
Product Brand:
Sigma-Aldrich
Product Number:
363782
Molecular Weight:
153.15
Molecular Formula:
CH3CONHC6H4F
CAS Number:
351-28-0
Test
Specification
Lot 06018EX Results
APPEARANCE:
WHITE TO YELLOW POWDER OR CRYSTALS
WHITE CRYSTALLINE POWDER
MELTING POINT:
82-84 DEGREES CELSIUS
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE
CONFORMS TO STRUCTURE
PROTON NMR SPECTRUM:
CONFORMS TO STRUCTURE
ELEMENTAL ANALYSIS:
CARBON 61.2%-64.3%
CARBON 62.37%
 :
NITROGEN 8.9%- 9.4%
HYDROGEN 5.25%
 :
NITROGEN 8.88%
THIN-LAYER:
CONSISTENT WITH 98% PURITY
CONSISTENT WITH 98% PURITY
SOLUBILITY:
25 MG/ML, METHANOL; CLEAR, COLORLESS TO
25 MG/ML, MEOH; CLEAR, COLORLESS SOLUTION
 :
APPROVED FEBRUARY 4, 2009 RJM
SEPTEMBER 2006: REASSAYED AND CONFIRMED THAT
 :
APPROVED FEBRUARY 4, 2009 RJM
MATERIAL CONTINUES TO MEET REQUIREMENTS.
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.