Certificate of Analysis
Sigma-Aldrich
Product Name:
2,4-Di-tert-amylphenol
Product Description:
99%
Product Brand:
Sigma-Aldrich
Product Number:
372099
Molecular Weight:
234.38
Molecular Formula:
[C2H5C(CH3)2]2C6H3OH
CAS Number:
120-95-6
Test
Specification
Lot 06606PX Results
APPEARANCE:
COLORLESS LIQUID OR WHITE SOLID
WHITE SOLID
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
CONFORMS TO STRUCTURE.
PROTON, CARBON NMR:
CONFORMS TO STRUCTURE.
GAS LIQUID:
98.5 % (MINIMUM)
99.7 %
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.