Certificate of Analysis
Sigma-Aldrich
Product Name:
Yttrium(III) isopropoxide solution
Product Description:
20-25 wt. % in toluene
Product Brand:
Sigma-Aldrich
Product Number:
380296
Molecular Weight:
266.17
Molecular Formula:
Y(OCH(CH3)2)3
CAS Number:
2172-12-5
Test
Specification
Lot 20706BE Results
APPEARANCE:
COLORLESS TO YELLOW LIQUID
COLORLESS LIQUID
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
CONFORMS TO STRUCTURE.
TITRATION:
7.7%-9.0% Y (COMPLEXOMETRIC)
8.9% Y (COMPLEXOMETRIC)
QUALITY CONTROL:
FEBRUARY 2006
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.