Certificate of Analysis
Sigma-Aldrich
Product Name:
2-Amino-7-chloro-5-oxo-5H-[1]benzopyrano[2,3-b]pyridine-3-carbonitrile
Product Description:
97%
Product Brand:
Sigma-Aldrich
Product Number:
402311
Molecular Weight:
271.66
CAS Number:
68302-10-3
Test
Specification
Lot 07515BG Results
APPEARANCE:
DULL YELLOW POWDER
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
PROTON, CARBON, APT:
CONFORMS TO STRUCTURE.
TITRATION:
101.0 % (WITH AGNO3 AFTER OXYGEN COMBUSTION)
HIGH PRESSURE LIQUID:
97.8 %
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.