Certificate of Analysis
Sigma-Aldrich
Product Name:
Di(ethylene glycol) vinyl ether
Product Description:
98%
Product Brand:
Sigma-Aldrich
Product Number:
409987
Molecular Weight:
132.16
Molecular Formula:
H2C=CH(OCH2CH2)2OH
CAS Number:
929-37-3
Test
Specification
Lot 09620JF Results
APPEARANCE:
YELLOW LIQUID
REFRACTIVE INDEX AT:
1.4428
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
PROTON, CARBON NMR:
CONFORMS TO STRUCTURE.
GAS LIQUID:
99.7%
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.