Certificate of Analysis
Sigma-Aldrich
Product Name:
Carbon-12C monoxide
Product Description:
99.95 atom % 12C
Product Brand:
Sigma-Aldrich
Product Number:
418072
Molecular Weight:
28.00
Molecular Formula:
12CO
CAS Number:
630-08-0
Test
Specification
Lot 10281AE Results
ISOTOPIC ASSAY:
99.96 ATOM % 12C
GAS LIQUID:
AR/O2 <20 PPM, CO2 = 29 PPM
CHROMATOGRAPHY:
N2 = 52 PPM, THC <20 PPM
 :
99.9 % PURITY
QUALITY CONTROL:
SEPTEMBER, 2004
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.