Certificate of Analysis
Sigma-Aldrich
Product Name:
4-Methyl-1-naphthaldehyde
Product Description:
97%
Product Brand:
Sigma-Aldrich
Product Number:
526487
Molecular Weight:
170.21
Molecular Formula:
CH3C10H6CHO
CAS Number:
33738-48-6
Test
Specification
Lot 11728MD Results
APPEARANCE:
WHITE TO YELLOW SOLID, POWDER OR CRYSTALS
FAINTT YELLOW SOLID
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
ELEMENTAL ANALYSIS:
CARBON 84.47%
GAS LIQUID:
96.5% (MINIMUM)
98.5%
QUALITY CONTROL:
NOVEMBER 2005
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.