Certificate of Analysis
Sigma-Aldrich
Product Name:
(4-Methoxyphenyl)diphenylsulfonium triflate
Product Brand:
Sigma-Aldrich
Product Number:
526967
Molecular Weight:
442.47
CAS Number:
116808-67-4
Test
Specification
Lot 06308CI Results
APPEARANCE:
WHITE CHUNKS
MELTING POINT:
101.0-103.0 DEGREES CELSIUS
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
PROTON NMR SPECTRUM:
CONFORMS TO STRUCTURE.
UV-VISIBLE SPECTRUM:
0.01002 G/L, H2O
 :
E259 = 14,352
ELEMENTAL ANALYSIS:
CARBON 54.36%
 :
HYDROGEN 3.75%
QUALITY CONTROL:
APRIL, 2000
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.