Certificate of Analysis
Sigma-Aldrich
Product Name:
PAMAM-25% C12 dendrimer, generation 2 solution
Product Description:
20 wt. % in methanol
Product Brand:
Sigma-Aldrich
Product Number:
536849
Molecular Weight:
4730.72
CAS Number:
204401-84-3
Test
Specification
Lot 02129AO Results
APPEARANCE:
COLORLESS LIQUID
REFRACTIVE INDEX AT:
1.3613
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
PROTON, CARBON NMR:
CONFORMS TO STANDARD. *
RESIDUE ON EVAPORATION:
22.16% SOLIDS
 :
* SUPPLIER DATA
QUALITY CONTROL:
APRIL 2001
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.