Certificate of Analysis
Sigma-Aldrich
Product Name:
Ethyl chlorofluoroacetate
Product Description:
98%
Product Brand:
Sigma-Aldrich
Product Number:
539147
Molecular Weight:
140.54
Molecular Formula:
Cl(F)CHCO2C2H5
CAS Number:
401-56-9
Test
Specification
Lot MKAA2927 Results
Appearance (Color):
Colorless to Faint Yellow
Colorless
Appearance (Form):
Liquid
Liquid
Infrared spectrum:
Conforms to Structure
Conforms
Titration by AgNO3:
97.5 - 102.5 %
100.0 %
 :
After Oxygen Combustion
Purity (GC):
≥97.5 %
99.7 %
Specification Date::
FEB 2009
Date of QC Release::
FEB 2009
Print Date::
FEB 27 2009
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.