Certificate of Analysis
Sigma-Aldrich
Product Name:
2,6-Difluorobenzenesulfonamide
Product Description:
97%
Product Brand:
Sigma-Aldrich
Product Number:
542709
Molecular Weight:
193.17
Molecular Formula:
F2C6H3SO2NH2
CAS Number:
60230-37-7
Test
Specification
Lot 06806TI Results
APPEARANCE:
WHITE TO BROWN POWDER, CRYSTALS, CRYSTALLINE
BEIGE POWDER
MELTING POINT:
187.2-191.3 DEGREES CELSIUS
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
CONFORMS TO STRUCTURE.
PROTON NMR SPECTRUM:
CONFORMS TO STRUCTURE.
ELEMENTAL ANALYSIS:
CARBON 36.0%-38.6%
CARBON 37.03%
 :
NITROGEN 6.9%- 7.5%
HYDROGEN 2.53%
 :
NITROGEN 7.23%
HIGH PRESSURE LIQUID:
96.5% (MINIMUM)
99.9 %
QUALITY CONTROL:
JANUARY 2001
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.