Certificate of Analysis
Sigma-Aldrich
Product Name:
4-Chlorobenzyl isocyanate
Product Description:
97%
Product Brand:
Sigma-Aldrich
Product Number:
571806
Molecular Weight:
167.59
Molecular Formula:
ClC6H4CH2NCO
CAS Number:
30280-44-5
Test
Specification
Lot 07922PD Results
APPEARANCE:
COLORLESS LIQUID
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
TITRATION:
100.5% (WITH AGNO3 AFTER OXYGEN COMBUSTION)
GAS LIQUID:
98.7%
QUALITY CONTROL:
NOVEMBER 2005
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.