Certificate of Analysis
Sigma-Aldrich
Product Name:
4,6-Dihydroxypyrimidine
Product Description:
98%
Product Brand:
Sigma-Aldrich
Product Number:
D120405
Molecular Weight:
112.09
CAS Number:
1193-24-4
Test
Specification
Lot 01231MI Results
APPEARANCE:
LIGHT ORANGE POWDER
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE AND STANDARD.
ELEMENTAL ANALYSIS:
CARBON 42.60%
 :
HYDROGEN 3.56%
 :
NITROGEN 24.38%
THIN-LAYER:
CONSISTENT WITH 98% PURITY
SOLUBILITY:
50MG/ML, 1N NH4OH; CLEAR, YELLOW SOLUTION
QUALITY CONTROL:
NOVEMBER, 2000
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.