Certificate of Analysis
Sigma-Aldrich
Product Name:
3,5-Dimethyl-1-pyrazolylformaminidium nitrate
Product Description:
97%
Product Brand:
Sigma-Aldrich
Product Number:
D182257
Molecular Weight:
201.18
CAS Number:
38184-47-3
Test
Specification
Lot 08201LE Results
APPEARANCE:
WHITE TO BEIGE POWDER
WHITE POWDER
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
CONFORMS TO STRUCTURE.
TITRATION:
96.5%-103.5% (WITH HCLO4)
100.5% (WITH HCLO4)
THIN-LAYER:
CONSISTENT WITH 97% PURITY
CONSISTENT WITH 97% PURITY
SOLUBILITY:
1G/10ML(100MG/ML), H2O; CLEAR TO VERY
1G/10ML, H2O; SLIGHTLY HAZY, VERY FAINT
 :
SLIGHTLY HAZY, COLORLESS TO FAINT YELLOW
YELLOW SOLUTION
QUALITY CONTROL:
SEPTEMBER 2006
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.