Certificate of Analysis
Sigma-Aldrich
Product Name:
Octadecanamide
Product Description:
technical grade, 85%
Product Brand:
Sigma-Aldrich
Product Number:
O601
Molecular Weight:
283.49
Molecular Formula:
CH3(CH2)16CONH2
CAS Number:
124-26-5
Test
Specification
Lot 04822MH Results
APPEARANCE:
WHITE TO BEIGE POWDER, FLAKES, CHIPS OR
OFF-WHITE BEADS
INFRARED SPECTRUM:
CONFORMS TO STRUCTURE.
ELEMENTAL ANALYSIS:
CARBON 64.0% - 88.4%
CARBON 75.87%
 :
NITROGEN 4.1% - 5.7%
HYDROGEN 13.16%
 :
NITROGEN 4.95%
GAS LIQUID:
84.0% (MINIMUM)
93.7%
PRODUCT CROSS:
REPLACES PRODUCT NUMBER 85704
QC RELEASE DATA:
OCTOBER 2007
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.