Certificate of Analysis
Sigma-Aldrich
Product Name:
Aluminum acetylacetonate
Product Description:
99% (titration)
Product Brand:
Sigma-Aldrich
Product Number:
A7955
Molecular Weight:
324.31
Molecular Formula:
Al(C5H7O2)3
CAS Number:
13963-57-0
Test
Specification
Lot 035H3633 Results
APPEARANCE:
WHITE POWDER WITH A LIGHT YELLOW CAST
SOLUBILITY:
CLEAR FAINT YELLOW SOLUTION AT 200 MG PLUS 4 ML OF 6 N HYDROCHLORIC ACID
IR SPECTRUM:
CONSISTENT WITH STRUCTURE
PURITY BY EDTA TITRATION:
8.38% ALUMINUM
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.