Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-CNOT3 antibody produced in rabbit
Product Description:
affinity isolated antibody
Product Brand:
Sigma-Aldrich
Product Number:
AV34318
Test
Specification
Lot QC3910 Results
Concentration:
0.5-1 mg/ml
0.5-1 mg/ml
ELISA:
Not Verified
Not Verified
Western Blotting:
Antibody dilution: 1 µg/ml
Antibody dilution: 1 µg/ml
Immunohistochemistry:
Not Verified
Not Verified
Immunofluorescence:
Not Verified
Not Verified
Immunoprecipitation:
Not Verified
Not Verified
Flow Cytometry:
Not Verified
Not Verified
Chromatin Immunoprecipitation:
Not Verified
Not Verified
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.