Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-MECP2 antibody produced in rabbit
Product Description:
Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Brand:
Sigma-Aldrich
Product Number:
HPA000593
Test
Specification
Lot A000786 Results
Protein Array:
pass
pass
Immunohistochemistry Dilution:
pass
1:600
Immunohistochemistry Antibody Retrieval Method:
pass
HIER pH6
Concentration:
0.09 mg/ml
Western Blot Score:
Band of predicted size in kDa (+/- 20%) with additional bands present
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.