Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-CELF2 antibody produced in rabbit
Product Description:
Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Brand:
Sigma-Aldrich
Product Number:
HPA035813
Test
Specification
Lot R33444 Results
Immunohistochemistry Dilution:
pass
1:200 - 1:500
Immunohistochemistry Dilution:
pass
1:200 - 1:500
Immunohistochemistry Dilution:
pass
1:200 - 1:500
Immunohistochemistry Dilution:
pass
1:200 - 1:500
Immunohistochemistry Antibody Retrieval Method:
pass
HIER pH6
Immunohistochemistry Antibody Retrieval Method:
pass
HIER pH6
Immunohistochemistry Antibody Retrieval Method:
pass
HIER pH6
Immunohistochemistry Antibody Retrieval Method:
pass
HIER pH6
Concentration:
pass
0.05
Concentration:
pass
0.05
Concentration:
pass
0.05
Concentration:
pass
0.05
Western Blot Score:
pass
Single band corresponding to predicted size in kDa (+/- 20%)
Western Blot Score:
pass
Single band corresponding to predicted size in kDa (+/- 20%)
Western Blot Score:
pass
Single band corresponding to predicted size in kDa (+/- 20%)
Western Blot Score:
pass
Single band corresponding to predicted size in kDa (+/- 20%)
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.