Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-THOC3 antibody produced in rabbit
Product Description:
Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Brand:
Sigma-Aldrich
Product Number:
HPA044009
Test
Specification
Lot R42293 Results
Protein Array:
pass
Immunohistochemistry Dilution:
1:125
Immunohistochemistry Antibody Retrieval Method:
HIER pH6
Concentration:
0,11
Western Blot Score:
Single band corresponding to predicted size in kDa (+/- 20%)
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.