Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-AKAP6 antibody produced in rabbit
Product Description:
Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Brand:
Sigma-Aldrich
Product Number:
HPA048741
Test
Specification
Lot R58683 Results
Immunohistochemistry Dilution:
Pass
1:1000 - 1:2500
Immunohistochemistry Antibody Retrieval Method:
Pass
HIER pH6
Concentration:
Report result
0.6 mg/mL
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.