Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-AMIGO2 antibody produced in rabbit
Product Description:
Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Product Brand:
Sigma-Aldrich
Product Number:
HPA059601
Test
Specification
Lot R84264 Results
Purification Method:
Affinity purified using the PrEST antigen as affinity ligand
Concentration:
0.2 mg/mL
Immunofluorescence fixation/permeabilization:
PFA/Triton X-100
Immunofluorescence working concentration:
1-4 ug/mL
Country of Origin:
Sweden
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.