Certificate of Analysis
Sigma-Aldrich
Product Name:
Butyl oleate
Product Description:
Grade I, ~99%
Product Brand:
Sigma-Aldrich
Product Number:
O7500
Molecular Weight:
338.57
CAS Number:
142-77-8
Test
Specification
Lot 116H8443 Results
APPEARANCE:
CLEAR COLORLESS LIQUID
SOLUBILITY:
CLEAR COLORLESS SOLUTION AT 50 MG/ML OF CHLOROFORM
PROTON NMR SPECTRUM:
CONSISTENT WITH STRUCTURE
PURITY BY GAS CHROMATOGRAPHY:
98.9%
PURITY BY THIN LAYER CHROMATOGRAPHY:
GREATER THAN 99%
QC RELEASE DATE:
DECEMBER 1996
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.