Certificate of Analysis
Sigma-Aldrich
Product Name:
Sodium acetate solution
Product Description:
0.3 M
Product Brand:
Sigma-Aldrich
Product Number:
S8388
Molecular Weight:
82.03
CAS Number:
127-09-3
Test
Specification
Lot 010K9010 Results
APPEARANCE:
CLEAR COLORLESS SOLUTION
PH TEST:
7.5
CONCENTRATION:
0.3 M (BY PERCHLORIC ACID TITRATION)
QC RELEASE DATE:
MARCH 2000
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.