Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-AMIGO2 antibody produced in mouse
Product Description:
IgG fraction of antiserum, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
SAB1400868
Test
Specification
Lot 09054 Results
Concentration:
0.1 mg/ml
Isotype:
not determined
Immunofluorescence (IF):
Antibody concentration: 10 ug/ml using HeLa cell.
pass
Western Blotting (Transfected lysate):
Antibody concentration: 0.1 ~ 5 ug/ml. Specific band of ~57.42 kDa using AMIGO2 transfected 293T lysate.
pass
Detection Antibody:
Suitable as a detection antibody in sandwich ELISA assay.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.