Certificate of Analysis
Sigma-Aldrich
Product Name:
Monoclonal Anti-VCP, (N-terminal) antibody produced in mouse
Product Description:
clone 2B2, purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
SAB1404523
Test
Specification
Lot 09170-2B2 Results
Concentration:
1 mg/mL
Isotype:
IgG1 Kappa
Western Blotting (Cell lysate):
Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~89.3 kDa using A-431 lysate.
pass
ELISA (Recombinant protein):
Antibody concentration: 1 ug/ml using recombinant protein.
pass
Western Blotting (Recombinant protein):
Antibody concentration: 1 ug/ml. Specific band of ~38.2 kDa using immunogen protein lysate.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.