Certificate of Analysis
Sigma-Aldrich
Product Name:
Monoclonal Anti-GNRHR2, (C-terminal) antibody produced in mouse
Product Description:
clone 1A5, purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
SAB1405341
Test
Specification
Lot 10096-1A5 Results
Concentration:
0.5 mg/mL
Isotype:
IgG1 Kappa
Sandwich ELISA:
Detection limit for recombinant GST tagged GNRHR2 is 0.03 ng/ml.
pass
ELISA (Recombinant protein):
Antibody concentration: 1 ug/ml using recombinant protein.
pass
Western Blotting (Recombinant protein):
Antibody concentration: 1 ug/ml. Specific band of ~32.3 kDa using immunogen protein lysate.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.