Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-SGSH antibody produced in mouse
Product Description:
purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
SAB1406433
Test
Specification
Lot 08303 Results
Concentration:
0.5 mg/mL
Isotype:
not determined
Western Blotting (Transfected lysate):
Antibody concentration: 0.1 ~ 5 ug/ml. Specific band of ~55.2 kDa using SGSH transfected 293T lysate.
pass
Detection Antibody:
Suitable as a detection antibody in sandwich ELISA assay.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.