Certificate of Analysis
Sigma-Aldrich
Product Name:
Anti-phospho-VCP (pSer352) antibody produced in rabbit
Product Description:
affinity isolated antibody
Product Brand:
Sigma-Aldrich
Product Number:
SAB4504738
Test
Specification
Lot 118449 Results
Appearance:
Colorless Liquid
Colorless Liquid
Concentration:
1 mg/mL
1 mg/mL
Immunogen:
Range AA337 to 367
Range AA337 to 367
ELISA:
1:1000
Pass
Western Blotting:
1:500~1:1000
Pass
Immunohistochemistry:
1:50~1:100
Pass
Immunofluorescence:
Not Verified
Not Verified
Immunoprecipitation:
Not Verified
Not Verified
Flow Cytometry:
Not Verified
Not Verified
Chromatin Immunoprecipitation:
Not Verified
Not Verified
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.