Certificate of Analysis
Sigma-Aldrich
Product Name:
Glyceryl tridodecanoate
Product Description:
~98%
Product Brand:
Sigma-Aldrich
Product Number:
T8641
Molecular Weight:
639.00
Molecular Formula:
[CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
CAS Number:
538-24-9
Test
Specification
Lot 128H0468 Results
APPEARANCE:
WHITE POWDER
SOLUBILITY:
CLEAR COLORLESS SOLUTION AT 250 MG PLUS 5.0 ML OF CHLOROFORM
PURITY BY THIN LAYER CHROMATOGRAPHY:
>99.5%
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.