Certificate of Analysis
Sigma-Aldrich
Product Name:
Urea solution
Product Description:
40 % (w/v) in H2O
Product Brand:
Sigma-Aldrich
Product Number:
U7129
Molecular Weight:
60.06
Molecular Formula:
NH2CONH2
CAS Number:
57-13-6
Test
Specification
Lot 090M1193 Results
Appearance (Color):
Colorless
Colorless
Appearance (Turbidity):
Clear
Clear
Appearance (Form):
Liquid
Liquid
pH:
7.80 - 8.20
8.20
Cultural Response:
Pass
Pass
 :
As described in data sheet
Specification Date::
AUG 2010
Date of QC Release::
SEP 2010
Print Date::
SEP 08 2010
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.