Certificate of Analysis
Sigma-Aldrich
Product Name:
Monoclonal Anti-CNOT3 antibody produced in mouse
Product Description:
clone 4B8, purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
WH0004849M1
Test
Specification
Lot 06139-4B8 Results
Concentration:
0.5 mg/ml
Isotype:
IgG1
Western Blotting (Cell lysate):
Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~82 kDa using Hela NE lysate.
pass
Immunofluorescence (IF):
Antibody concentration: 10 ug/ml using HeLa cell.
pass
ELISA (Recombinant protein):
Antibody concentration: 1 ug/ml using recombinant protein.
pass
Western Blotting (Recombinant protein):
Antibody concentration: 1 ug/ml. Specific band of ~37 kDa using immunogen protein lysate.
pass
Western Blotting (Transfected lysate):
Antibody concentration: 0.1 ~ 5 ug/ml. Specific band of ~82 kDa using CNOT3 transfected 293T lysate.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.