Certificate of Analysis
Sigma-Aldrich
Product Name:
Monoclonal Anti-PTK2 antibody produced in mouse
Product Description:
clone 2C3, purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
WH0005747M1
Test
Specification
Lot 06090-2C3 Results
Concentration:
1 mg/ml
Isotype:
IgG1
Western Blotting (Cell lysate):
Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~119 kDa using Hela NE lysate.
pass
Sandwich ELISA:
Detection limit for recombinant GST tagged PTK2 is approximately 0.03ng/ml.
pass
ELISA (Recombinant protein):
Antibody concentration: 1 ug/ml using recombinant protein.
pass
Western Blotting (Recombinant protein):
Antibody concentration: 1 ug/ml. Specific band of ~41 kDa using immunogen protein lysate.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.