Certificate of Analysis
Sigma-Aldrich
Product Name:
Monoclonal Anti-RORA antibody produced in mouse
Product Description:
clone 4E3, purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
WH0006095M1
Test
Specification
Lot 05347-4E3 Results
Concentration:
1 mg/ml
Isotype:
IgG
Sandwich ELISA:
Detection limit for recombinant GST tagged RORA is approximately 0.3ng/ml.
pass
ELISA (Recombinant protein):
Antibody concentration: 1 ug/ml using recombinant protein.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.