Certificate of Analysis
Sigma-Aldrich
Product Name:
Monoclonal Anti-LAT antibody produced in mouse
Product Description:
clone 3B11-1D5, purified immunoglobulin, buffered aqueous solution
Product Brand:
Sigma-Aldrich
Product Number:
WH0027040M1
Test
Specification
Lot 05285-S1 Results
Concentration:
1 mg/ml
Isotype:
IgG1
Western Blotting (Cell lysate):
Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~28 kDa using HL-60 lysate.
pass
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P):
Antibody concentration: 5 ug/ml using human tonsil tissue.
pass
Sandwich ELISA:
Detection limit for recombinant GST tagged LAT is approximately 0.03ng/ml.
pass
ELISA (Recombinant protein):
Antibody concentration: 1 ug/ml using recombinant protein.
pass
Western Blotting (Recombinant protein):
Antibody concentration: 1 ug/ml. Specific band of ~51 kDa using immunogen protein lysate.
pass
Tom Beil, VP, Quality and Regulatory Affairs
The above information is believed to be correct to the best of our knowledge. This information is to be used for the purpose of determining animal origin only and not to be confused with "country of origin" for import/export purposes. Sigma-Aldrich shall not be held liable for any damage resulting from handling or from contact with the above product.