Breaking the Boundaries of Biomolecule Separation with BIOshell™ UHPLC Columns
WEBINAR
Superficially porous particles (SPPs) have advantages over traditional fully porous particles (FPPs) when packed into columns for chromatographic separation of biomolecules. Due to the presence of a solid silica core, analytes have a smaller distance to travel within a particle, thus minimizing diffusion due to mass transfer effects. Combined with a large pore diameter and a thin, porous shell, these particles can yield highly efficient peaks even for large proteins such as monoclonal antibodies.
In this webinar, we will discuss:
In this webinar, you will learn about the advantages of using SPP-packed BIOshell™ columns for various large molecule separation challenges. Aspects of peptide mapping, intact analysis and middle-up analysis will be reviewed, and strategies for improving these assays will be provided. In addition, a unique 1.5 mm I.D. column geometry and its advantages in improving sensitivity and making methods more sustainable will be reviewed. Finally, strategies for characterizing oligonucleotides will be discussed in relation to how BIOshell™ can assist in analyzing this class of biomolecule.
Key Learning Objectives
- Discover the importance of SPP particle architecture (pore size and shell thickness) and how it contributes to high-efficiency separations of biomolecules
- Explore how column chemistry can play a role in improving peptide mapping accuracy
- Learn how extra column dispersion can affect chromatographic performance and discover strategies to mitigate it
Speakers
Cory E. Muraco
MilliporeSigma
Senior Global Product Manager, Large Molecule HPLC Columns
Cory E. Muraco is the Senior Global Product Manager for Large Molecule HPLC Columns at MilliporeSigma, the life science business of Merck KGaA, Darmstadt, Germany. After finishing his graduate studies at Youngstown State University (Youngstown, Ohio, USA), Cory started his career at MilliporeSigma in 2013. After holding several roles in both R&D and Marketing functions across the past decade, in 2026, Cory assumed his current role as the Large Molecule HPLC Columns Product Manager where he is tasked with designing, developing, and executing the marketing and R&D strategies around MilliporeSigma’s biomolecule HPLC columns. Cory is the author of several manuscripts appearing in trade magazines and has delivered over 100 presentations at international conferences, round table symposia, and at various pharmaceutical and biopharmaceutical companies.
Pharma and biopharma manufacturing
- Pharma and biopharma manufacturing
Duration:1h
Language:English
Session 1:presented June 30, 2023